1-(4-Bromophenyl)heptan-1-ol ≥97%

Supplier: AOBChem USA

Warning

129678-1G 129678-25G 129678-500MG 129678-5G
76882-568EA 233.4 USD
76882-568 76882-570 76882-572 76882-574
1-(4-Bromophenyl)heptan-1-ol ≥97%
1-(4-Bromophenyl)heptan-1-ol
Formula: C₁₃H₁₉BrO
MW: 271.193 g/mol
Storage Temperature: Refrigerator
CAS Number: 71434-33-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR