(5-Fluoro-3-pyridyl)methanol ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
(5-Fluoropyridin-3-yl)methanol
12917-1G
12917-25G
12917-5G
76882-544EA
62.6
USD
76882-544
76882-546
76882-548
(5-Fluoro-3-pyridyl)methanol ≥97%
(5-Fluoro-3-pyridyl)methanol
Formula:
C₆H₆FNO MW: 127.12 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD01758680 CAS Number: 22620-32-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...