(3-Chlorophenyl)(furan-2-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

128305-1G 128305-25G 128305-500MG 128305-5G
76882-528EA 350.1 USD
76882-528 76882-530 76882-532 76882-534
(3-Chlorophenyl)(furan-2-yl)methanol ≥97%
(3-Chlorophenyl)(furan-2-yl)methanol
Formula: C₁₁H₉ClO₂
Storage Temperature: Refrigerator
CAS Number: 944652-59-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR