(3,5-Difluorophenyl)(Phenyl)Sulfane ≥97% research grade

Supplier: AOBChem USA

Warning

127499-1G 127499-25G 127499-500MG 127499-5G
76882-490EA 318.27 USD
76882-490 76882-492 76882-494 76882-496
(3,5-Difluorophenyl)(Phenyl)Sulfane ≥97% research grade
(3,5-Difluorophenyl)(Phenyl)Sulfane
Formula: C₁₂H₈F₂S
Storage Temperature: Ambient
CAS Number: 1632130-37-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR