(2-Bromo-4-chloro-5-methoxyphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

126908-1G 126908-500MG
76882-484EA 1891.59 USD
76882-484 76882-486
(2-Bromo-4-chloro-5-methoxyphenyl)boronic acid ≥97%
(2-Bromo-4-chloro-5-methoxyphenyl)boronic acid
Formula: C₇H₇BBrClO₃
MW: 265.297 g/mol
Storage Temperature: Ambient
CAS Number: 2225176-63-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR