2',3',6'-Trifluoroacetophenone ≥97%

Supplier: AOBChem USA

Warning

12629-25G 12629-5G
76882-476EA 268.41 USD
76882-476 76882-478
2',3',6'-Trifluoroacetophenone ≥97%
2',3',6'-Trifluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₅F₃O
Storage Temperature: Ambient
MDL Number: MFCD00070809
CAS Number: 208173-22-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR