2,3,5-Trifluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

12556-25G 12556-5G
76882-456EA 177.18 USD
76882-456 76882-458
2,3,5-Trifluorophenylboronic acid ≥97%
2,3,5-Trifluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₄BF₃O₂
MW: 175.9 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01863166
CAS Number: 247564-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR