2,3,4-Trichlorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

12544-25G 12544-5G
76882-446EA 346.92 USD
76882-446 76882-448
2,3,4-Trichlorophenylboronic acid ≥97%
2,3,4-Trichlorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₄BCl₃O₂
Storage Temperature: Ambient
MDL Number: MFCD03095112
CAS Number: 352530-21-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR