2,4-Dibromo-m-toluidine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,4-Dibromo-3-methylaniline

123233-1G 123233-25G 123233-500MG 123233-5G
76882-408EA 274.78 USD
76882-408 76882-410 76882-412 76882-414
2,4-Dibromo-m-toluidine ≥97%
2,4-Dibromo-m-toluidine
Formula: C₇H₇Br₂N
Storage Temperature: Ambient
MDL Number: MFCD12547774
CAS Number: 80948-78-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR