2-(Methylthio)phenylboronic acid pinacolate ≥97%

Supplier: AOBChem USA

Warning

12146-25G 12146-5G
76882-360EA 198.39 USD
76882-360 76882-362
2-(Methylthio)phenylboronic acid pinacolate ≥97%
2-(Methylthio)phenylboronic acid pinacolate
Formula: C₁₃H₁₉BO₂S
Storage Temperature: Ambient
MDL Number: MFCD05155219
CAS Number: 1072945-09-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR