1-Bromo-2-Methyl-3-(Methylsulfonyl)Benzene ≥97%
Supplier: AOBChem USA
|
Warning
|
120646-1G
120646-25G
120646-500MG
120646-5G
76882-348EA
287.51
USD
76882-348
76882-350
76882-352
76882-354
1-Bromo-2-Methyl-3-(Methylsulfonyl)Benzene ≥97%
1-Bromo-2-Methyl-3-(Methylsulfonyl)Benzene
Formula:
C₈H₉BrO₂S MW: 249.125 g/mol Storage Temperature: Ambient |
CAS Number:
871352-94-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...