(2,4-Dichloro-5-(Trifluoromethyl)Phenyl)(Methyl)Sulfane ≥97%

Supplier: AOBChem USA

Warning

119636-1G 119636-500MG 119636-5G 119636-25G
76882-322EA 2205.62 USD
76882-322 76882-324 77631-514 77657-274
(2,4-Dichloro-5-(Trifluoromethyl)Phenyl)(Methyl)Sulfane ≥97%
(2,4-Dichloro-5-(Trifluoromethyl)Phenyl)(Methyl)Sulfane

Precaución: for Research Use Only

Formula: C₈H₅Cl₂F₃S
MW: 261.092 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34784466
CAS Number: 2709895-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR