2,3,6-Trifluorophenol ≥97%

Supplier: AOBChem USA

Warning

11860-25G 11860-5G
76882-314EA 204.76 USD
76882-314 76882-316
2,3,6-Trifluorophenol ≥97%
2,3,6-Trifluorophenol

Precaución: for Research Use Only

Formula: C₆H₃F₃O
Storage Temperature: Ambient
MDL Number: MFCD00061214
CAS Number: 113798-74-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR