2,3-Dichloro-4-pyridineboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,3-Dichloropyridine-4-boronic acid, 2,3-Dichloro-4-pyridylboronic acid

11541-25G 11541-5G
76882-224EA 268.41 USD
76882-224 76882-226
2,3-Dichloro-4-pyridineboronic acid ≥97%
2,3-Dichloro-4-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₅H₄BCl₂NO₂
MW: 191.81 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04038756
CAS Number: 951677-39-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR