2,3-Dibromo-6-picoline ≥97%
Supplier: AOBChem USA
|
Warning
|
11531-25G
11531-5G
76882-218EA
204.76
USD
76882-218
76882-220
2,3-Dibromo-6-picoline ≥97%
2,3-Dibromo-6-picoline
Precaución: for Research Use Only
Formula:
C₆H₅Br₂N Storage Temperature: Ambient |
MDL Number:
MFCD03095214 CAS Number: 261373-04-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...