2-(Cyclopropylmethoxy)phenylboronic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
11458-1G
11458-25G
11458-500MG
11458-5G
76882-204EA
129.43
USD
76882-204
76882-206
76882-208
76882-210
2-(Cyclopropylmethoxy)phenylboronic acid ≥97%
2-(Cyclopropylmethoxy)phenylboronic acid
Precaución: for Research Use Only
Formula:
C₁₀H₁₃BO₃ Storage Temperature: Ambient |
MDL Number:
MFCD04039015 CAS Number: 1050510-36-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...