2-[(4-Methylbenzyl)oxy]benzoic acid ≥97%

Supplier: AOBChem USA

Warning

114253-1G 114253-25G 114253-500MG 114253-5G
76882-190EA 130.5 USD
76882-190 76882-192 76882-194 76882-196
2-[(4-Methylbenzyl)oxy]benzoic acid ≥97%
2-[(4-Methylbenzyl)oxy]benzoic acid
Formula: C₁₅H₁₄O₃
Storage Temperature: Ambient
MDL Number: MFCD09719413
CAS Number: 52803-45-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR