1-Bromo-2-((4-methylbenzyl)oxy)benzene ≥97% research grade

Supplier: AOBChem USA

Warning

114220-1G 114220-25G 114220-500MG 114220-5G
76882-174EA 349.04 USD
76882-174 76882-176 76882-178 76882-180
1-Bromo-2-((4-methylbenzyl)oxy)benzene ≥97% research grade
1-Bromo-2-((4-methylbenzyl)oxy)benzene
Formula: C₁₄H₁₃BrO
Storage Temperature: Ambient
CAS Number: 1036452-97-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR