1-Chloro-3-(methoxymethoxy)benzene 95%

Supplier: AOBChem USA

Warning

11168-1G 11168-25G 11168-5G 11168-500MG
76882-144EA 97.61 USD
76882-144 76882-146 76882-148 77648-348
1-Chloro-3-(methoxymethoxy)benzene 95%
1-Chloro-3-(methoxymethoxy)benzene

Precaución: for Research Use Only

Formula: C₈H₉ClO₂
MW: 172.609 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18311888
CAS Number: 91105-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR