2-(3-Bromophenyl)-2-methyl-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

11118-1G 11118-25G 11118-5G
76882-114EA 62.6 USD
76882-114 76882-116 76882-118
2-(3-Bromophenyl)-2-methyl-1,3-dioxolane ≥97%
2-(3-Bromophenyl)-2-methyl-1,3-dioxolane
Formula: C₁₀H₁₁BrO₂
MW: 243.097 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09831921
CAS Number: 39172-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR