2-(4-Bromophenyl)-1,3-dioxolane ≥95%

Supplier: AOBChem USA

Warning

10398-25G 10398-5G
76882-046EA 177.18 USD
76882-046 76882-048
2-(4-Bromophenyl)-1,3-dioxolane ≥95%
2-(4-Bromophenyl)-1,3-dioxolane
Formula: C₉H₉BrO₂
Storage Temperature: Refrigerator
MDL Number: MFCD01318950
CAS Number: 10602-01-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR