1-(5-Bromo-2-fluoro-3-methylphenyl)-4,4-dimethylcyclohexanol 95%

Supplier: AOBChem USA

Warning

102609-1G 102609-500MG 102609-5G 102609-25G
76882-038EA 527.27 USD
76882-038 76882-040 76882-042 77653-150
1-(5-Bromo-2-fluoro-3-methylphenyl)-4,4-dimethylcyclohexanol 95%
1-(5-Bromo-2-fluoro-3-methylphenyl)-4,4-dimethylcyclohexanol

Precaución: for Research Use Only

Formula: C₁₅H₂₀BrFO
MW: 315.221 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD34777196
CAS Number: 2918923-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR