2-(3-Methyl-2-(methylthio)phenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

101868-1G 101868-25G 101868-500MG 101868-5G
76882-024EA 271.6 USD
76882-024 76882-026 76882-028 76882-030
2-(3-Methyl-2-(methylthio)phenyl)-1,3-dioxolane ≥97%
2-(3-Methyl-2-(methylthio)phenyl)-1,3-dioxolane
Formula: C₁₁H₁₄O₂S
MW: 210.293 g/mol
Storage Temperature: Refrigerator
CAS Number: 2918919-58-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR