5-Bromo-N,4-dimethylnicotinamide

Supplier: AOBChem USA

Warning

45755-1G 45755-25G 45755-500MG 45755-5G
76932-532EA 350.1 USD
76932-532 76932-534 76932-536 76932-538
5-Bromo-N,4-dimethylnicotinamide
5-Bromo-N,4-dimethylnicotinamide
Formula: C₈H₉BrN₂O
MW: 229.074 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403810
CAS Number: 2246722-11-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR