(2,4,5-Trimethoxyphenyl)boronic acid research grade

Supplier: AOBChem USA

Warning

54096-1G 54096-25G 54096-500MG 54096-5G
76891-544EA 439.22 USD
76891-544 76891-546 76891-548 76891-550
(2,4,5-Trimethoxyphenyl)boronic acid research grade
(2,4,5-Trimethoxyphenyl)boronic acid
Formula: C₉H₁₃BO₅
MW: 212.008 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11226499
CAS Number: 213698-56-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR