6-Chloro-2-fluoro-3-methoxybenzamide ≥97% research grade

Supplier: AOBChem USA

Warning

55445-25G 55445-5G
76943-228EA 616.39 USD
76943-228 76943-230
6-Chloro-2-fluoro-3-methoxybenzamide ≥97% research grade
6-Chloro-2-fluoro-3-methoxybenzamide
Formula: C₈H₇ClFNO₂
Storage Temperature: Ambient
MDL Number: MFCD06660354
CAS Number: 886761-75-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR