5-Phenyl-1,3-thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Phenylthiazol-2-amine, 2-Amino-5-phenylthiazole, 5-Phenyl-1,3-thiazol-2-amine, 5-Phenyl-2-thiazolamine

54965-1G 54965-25G 54965-5G
76943-200EA 62.6 USD
76943-200 76943-202 76943-204
5-Phenyl-1,3-thiazol-2-amine ≥97%
5-Phenyl-1,3-thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₈N₂S
Storage Temperature: Ambient
MDL Number: MFCD00022451
CAS Number: 39136-63-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR