N'-(4-Methoxybenzylidene)-4-Methylbenzenesulfonohydrazide ≥97%

Supplier: AOBChem USA

Warning

46538-1G 46538-25G 46538-500MG 46538-5G
76942-630EA 144.29 USD
76942-630 76942-632 76942-634 76942-636
N'-(4-Methoxybenzylidene)-4-Methylbenzenesulfonohydrazide ≥97%
N'-(4-Methoxybenzylidene)-4-Methylbenzenesulfonohydrazide
Formula: C₁₅H₁₆N₂O₃S
MW: 304.364 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00412025
CAS Number: 19350-72-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR