tert-Butyl (2-bromo-5-methylphenyl)carbamate ≥97% research grade

Supplier: AOBChem USA

Warning

38248-1G 38248-25G 38248-5G
76942-148EA 117.76 USD
76942-148 76942-150 76942-152
tert-Butyl (2-bromo-5-methylphenyl)carbamate ≥97% research grade
tert-Butyl (2-bromo-5-methylphenyl)carbamate
Formula: C₁₂H₁₆BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD24229813
CAS Number: 1186637-29-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR