6-Bromo-8-methyl-[1,2,4]triazolo[4,3-a]pyridine ≥97%

Supplier: AOBChem USA

Warning

36857-1G 36857-25G 36857-500MG 36857-5G
76941-604EA 350.1 USD
76941-604 76941-606 76941-608 76941-610
6-Bromo-8-methyl-[1,2,4]triazolo[4,3-a]pyridine ≥97%
6-Bromo-8-methyl-[1,2,4]triazolo[4,3-a]pyridine
Formula: C₇H₆BrN₃
MW: 212.047 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14551607
CAS Number: 1216301-85-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR