2-Chloro-3-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-Chloro-5-methoxypyridine-3-boronic acid pinacol ester

31673-1G 31673-25G 31673-500MG 31673-5G
76939-992EA 264.17 USD
76939-992 76939-994 76939-996 76939-998
2-Chloro-3-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine ≥97%
2-Chloro-3-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Formula: C₁₂H₁₇BClNO₃
MW: 269.53 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16660309
CAS Number: 1256360-28-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR