Methyl-6-bromobenzo[b]thiophene-2-carboxylate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Methyl 6-bromo-1-benzothiophene-2-carboxylate, Methyl 6-bromobenzo[b]thiophene-2-carboxylate

26418-25G 26418-5G
76938-874EA 240.83 USD
76938-874 76938-876
Methyl-6-bromobenzo[b]thiophene-2-carboxylate ≥97%
Methyl 6-bromobenzo[b]thiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₇BrO₂S
MW: 271.1300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07371541
CAS Number: 360576-01-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR