Methyl 2-fluoro-5-methylpyridine-3-carboxylate ≥97%

Supplier: AOBChem USA

Warning

23056-1G 23056-25G 23056-500MG 23056-5G
76938-208EA 379.81 USD
76938-208 76938-210 76938-212 76938-214
Methyl 2-fluoro-5-methylpyridine-3-carboxylate ≥97%
Methyl 2-fluoro-5-methylpyridine-3-carboxylate
Formula: C₈H₈FNO₂
MW: 169.153 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16611612
CAS Number: 1227575-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR