6-Bromo-3-chloro-2-fluorophenol ≥97%

Supplier: AOBChem USA

Warning

18824-1G 18824-25G 18824-500MG 18824-5G
76937-378EA 155.96 USD
76937-378 76937-380 76937-382 76937-384
6-Bromo-3-chloro-2-fluorophenol ≥97%
6-Bromo-3-chloro-2-fluorophenol
Formula: C₆H₃BrClFO
Storage Temperature: Refrigerator
MDL Number: MFCD19105360
CAS Number: 186590-40-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR