4-Iodo-1,2-dihydropyridin-2-one ≥97%

Supplier: AOBChem USA

Warning

50869-1G 50869-25G 50869-5G
76933-000EA 84.88 USD
76933-000 76933-002 76933-004
4-Iodo-1,2-dihydropyridin-2-one ≥97%
4-Iodo-1,2-dihydropyridin-2-one
Formula: C₅H₄INO
Storage Temperature: Refrigerator
MDL Number: MFCD09702419
CAS Number: 858839-90-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR