5-Bromo-2-(2-methoxyethoxy)pyrimidine ≥97%

Supplier: AOBChem USA

Warning

48675-25G 48675-5G
76932-828EA 453.01 USD
76932-828 76932-830
5-Bromo-2-(2-methoxyethoxy)pyrimidine ≥97%
5-Bromo-2-(2-methoxyethoxy)pyrimidine
Formula: C₇H₉BrN₂O₂
MW: 233.063 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18903069
CAS Number: 1289131-96-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR