5-Bromo-N-(tert-butyl)pyrimidin-2-amine ≥97%

Supplier: AOBChem USA

Warning

45672-25G 45672-5G
76932-518EA 346.92 USD
76932-518 76932-520
5-Bromo-N-(tert-butyl)pyrimidin-2-amine ≥97%
5-Bromo-N-(tert-butyl)pyrimidin-2-amine
Formula: C₈H₁₂BrN₃
MW: 230.105 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12546490
CAS Number: 14001-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR