4-Fluoro-1-isopropoxy-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

36751-25G 36751-5G
76931-890EA 524.09 USD
76931-890 76931-892
4-Fluoro-1-isopropoxy-2-methylbenzene ≥97%
4-Fluoro-1-isopropoxy-2-methylbenzene
Formula: C₁₀H₁₃FO
MW: 168.208 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23114432
CAS Number: 1369909-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR