4-Chloro-3-fluorobenzotrifluoride ≥97%
Supplier: AOBChem USA
|
Warning
|
27757-25G
27757-5G
76929-628EA
222.79
USD
76929-628
76929-630
4-Chloro-3-fluorobenzotrifluoride ≥97%
4-Chloro-3-fluorobenzotrifluoride
Precaución: for Research Use Only
Formula:
C₇H₃ClF₄ MW: 198.55 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00060646 CAS Number: 32137-20-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...