5-Bromo-2-(methoxymethyl)pyridine ≥97%
Supplier: AOBChem USA
|
Warning
|
25656-1G
25656-500MG
25656-5G
76928-856EA
647.15
USD
76928-856
76928-858
76928-860
5-Bromo-2-(methoxymethyl)pyridine ≥97%
5-Bromo-2-(methoxymethyl)pyridine
Formula:
C₇H₈BrNO MW: 202.049 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD18254265 CAS Number: 1000787-43-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...