5-Fluoro-3-(methylthio)benzyl alcohol ≥97% research grade
Supplier: AOBChem USA
|
Warning
|
24559-1G
24559-25G
24559-500MG
24559-5G
76928-438EA
290.69
USD
76928-438
76928-440
76928-442
76928-444
5-Fluoro-3-(methylthio)benzyl alcohol ≥97% research grade
5-Fluoro-3-(methylthio)benzyl alcohol
Formula:
C₈H₉FOS Storage Temperature: Ambient |
MDL Number:
MFCD28142583 CAS Number: 851785-58-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...