4-Ethoxy-2,6-difluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

21096-25G 21096-5G
76927-340EA 304.48 USD
76927-340 76927-342
4-Ethoxy-2,6-difluorobenzaldehyde ≥97%
4-Ethoxy-2,6-difluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₈F₂O₂
MW: 186.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09258695
CAS Number: 1017779-48-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR