(5-Fluoro-3-formyl-2-methoxyphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Fluoro-3-formyl-2-methoxyphenylboronic acid

17321-1G 17321-25G 17321-500MG 17321-5G
76926-154EA 153.84 USD
76926-154 76926-156 76926-158 76926-160
(5-Fluoro-3-formyl-2-methoxyphenyl)boronic acid ≥97%
(5-Fluoro-3-formyl-2-methoxyphenyl)boronic acid
Formula: C₈H₈BFO₄
MW: 197.96 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09265144
CAS Number: 1072951-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR