4-Borono-3-fluorobenzoic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 4-Carboxy-2-fluorophenylboronic acid

10028-25G 10028-5G
76924-184EA 304.48 USD
76924-184 76924-186
4-Borono-3-fluorobenzoic acid ≥97% research grade
4-Carboxy-2-fluorophenylboronic acid
Formula: C₇H₆BFO₄
Storage Temperature: Ambient
MDL Number: MFCD08458473
CAS Number: 851335-07-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR