2-Bromo-4-ethyl-1-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

57602-1G 57602-25G 57602-5G
76905-276EA 64.72 USD
76905-276 76905-278 76905-280
2-Bromo-4-ethyl-1-methoxybenzene ≥97%
2-Bromo-4-ethyl-1-methoxybenzene
Formula: C₉H₁₁BrO
Storage Temperature: Ambient
MDL Number: MFCD18783172
CAS Number: 99179-98-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR