2-Fluoro-1-methoxy-4-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

51369-25G 51369-5G
76904-662EA 531.52 USD
76904-662 76904-664
2-Fluoro-1-methoxy-4-(trifluoromethyl)benzene ≥97%
2-Fluoro-1-methoxy-4-(trifluoromethyl)benzene
Formula: C₈H₆F₄O
MW: 194.126 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13185638
CAS Number: 78263-49-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR