2-Methoxy-4-methylnicotinaldehyde 95%

Supplier: AOBChem USA

Warning

36516-1G 36516-500MG 36516-5G 36516-25G
76903-292EA 473.17 USD
76903-292 76903-294 76903-296 77655-414
2-Methoxy-4-methylnicotinaldehyde 95%
2-Methoxy-4-methylnicotinaldehyde
Formula: C₈H₉NO₂
MW: 151.163 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16611506
CAS Number: 123506-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR