2-Chloro-4-iodo-5-methoxypyridine ≥97%
Supplier: AOBChem USA
|
Warning
|
35365-1G
35365-500MG
35365-5G
76903-112EA
471.04
USD
76903-112
76903-114
76903-116
2-Chloro-4-iodo-5-methoxypyridine ≥97%
2-Chloro-4-iodo-5-methoxypyridine
Formula:
C₆H₅ClINO Storage Temperature: Refrigerator |
MDL Number:
MFCD16610524 CAS Number: 1211516-07-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...