2,6-Dimethyl-4-chlorophenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

33706-1G 33706-25G 33706-500MG 33706-5G
76902-676EA 263.11 USD
76902-676 76902-678 76902-680 76902-682
2,6-Dimethyl-4-chlorophenylboronic acid pinacol ester ≥97%
2,6-Dimethyl-4-chlorophenylboronic acid pinacol ester
Formula: C₁₄H₂₀BClO₂
MW: 266.571 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD23380471
CAS Number: 1374578-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR