2-Methylbenzo[b]thiophene ≥97%

Supplier: AOBChem USA

Warning

32073-25G 32073-5G
76902-168EA 559.1 USD
76902-168 76902-170
2-Methylbenzo[b]thiophene ≥97%
2-Methylbenzo[b]thiophene

Precaución: for Research Use Only

Formula: C₉H₈S
MW: 148.23 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00216250
CAS Number: 1195-14-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR